Imprimer
PIG® mini drip deck
Fournisseur: NEW PIG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PAK371 PAK372
121-0535EA 60.4 EUR
121-0535 121-0536
PIG® mini drip deck
Plateaux/Récipients/Pots Plateaux de déversements
If containers leak, drip or crack, make sure a Mini Drip Deck with built-in sump is in position to catch the liquid.
  • Translucent sidewalls let you easily inspect fluid level in sump
  • Compact size fits on tabletop or counter; ideal for use in labs
  • Pour spout for easy draining
Chemical-resistant polyethylene tray features built-in sump containment to trap leaks and drips whilst you work.
Attention: If using this product with flammable liquids, please consider the regulations that apply to storage and handling of flammable liquids and the safety of this application, specifically flammable vapours, static discharge and heat sources.