Imprimer
Irradiated cleanroom tapes, CR100IR™
Fournisseur: MICRONOVA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
CR100-34WHIR CR100-1WHIR CR100-12YEIR CR100-34YEIR CR100-1TRIR CR100-34TRIR CR100-2REIR CR10034WHIRGSK CR100-2TRIR CR100-2WHIR CR100-1LBIR CR10034WHIR
817-0067EA 45.7 EUR
817-0067 819-0019 819-0087 819-0093 819-0088 819-0091 MICRCR100-2REIR MICRCR10034WHIRGSK MICRCR100-2TRIR MICRCR100-2WHIR MICRCR100-1LBIR MICRCR10034WHIR
Irradiated cleanroom tapes, CR100IR™
Tapes
These ultra-processed irradiated cleanroom tapes are made of vinyl and are processed in a ISO 4 environment.
  • Suitable for critical and aseptic processing applications, media preparation, batching, packaging within the laboratory or sterile processing area
  • Helps to limit particulate and bioburden counts
  • Extremely conformable; tapes and seals curved and irregular surfaces
  • Clean peel from wafer cartridges and cleanroom work surfaces
  • Matte finish can be written on with a cleanroom pen
  • Wound on a 7.6 cm (3”) plastic core
The tape comes with full certification of irradiation and lot traceability.
Length: 33 m
Different colours available.
Cdt: Individually double bagged.