Imprimer
Electrolyte solutions, organic
Marques : SI Analytics
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
285138308 285138324
SCOI285138308EA 104 EUR
SCOI285138308 SCOI285138324
Electrolyte solutions, organic
Électrolytes Solutions de remplissage d'électrode
Electrolyte solutions for measurements in organic solutions for reference electrodes and as electrolyte bridges.