Imprimer

Anti-E Tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Fournisseur: Bioss
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BOSSBS-2116R-HRPEA 347 EUR
BOSSBS-2116R-HRP
Anti-E Tag Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorps
E Tag is a commonly used epitope tag engineered onto the N- or C- terminus of a protein of interest so that the tagged protein can be analyzed and visualized. Because of the small size of the epitope, it is unlikely to alter the activity of the cloned sequence.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: E Tag
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Tags
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR