Imprimer

Anti-CALB2 Rabbit Polyclonal Antibody (Cy3®)

Fournisseur: Bioss
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

BOSSBS-0062R-CY3EA 356 EUR
BOSSBS-0062R-CY3
Anti-CALB2 Rabbit Polyclonal Antibody (Cy3®)
Anticorps
Calretinin is a calcium-binding protein which is abundant in auditory neurons. It belongs to the calbindin family. Calbindin 2 (calretinin), closely related to calbindin 1, is an intracellular calcium-binding protein belonging to the troponin C superfamily. Calbindin 1 is known to be involved in the vitamin-D-dependent calcium absorption through intestinal and renal epithelia, while the function of neuronal calbindin 1 and calbindin2 is poorly understood. The sequence of the calbindin 2 cDNA reveals an open reading frame of 271 codons coding for a protein of 31,520 Da, and shares 58% identical residues with human calbindin1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CALB2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR