Imprimer
Cation exchange chromatography columns, TSK-GEL® CM STAT
Fournisseur: TOSOH Bioscience
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0021965 0021966
TOYO0021965EA 1320 EUR
TOYO0021965 TOYO0021966
Cation exchange chromatography columns, TSK-GEL® CM STAT
Colonnes pour chromatographie
TSK-GEL® CM STAT are cross-linked hydrophilic polymer (mono-disperse particles) based columns designed to analyse the profile of charge isomers of proteins. These STAT columns provide fast, high resolution separations at moderate pressures.
  • Nonporous columns
  • High efficiency separation at short analysis time
  • Used for high resolution protein separation