Imprimer
Cleaning cloths for glass and smooth surfaces, Clino®
Fournisseur: PFENNIG REINIGUNGSTECHNIK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2650002 2650004
PPSA2650002EA 318 EUR
PPSA2650002 PPSA2650004
Cleaning cloths for glass and smooth surfaces, Clino®
Lingettes Chiffons nettoyants réutilisables Chiffons pour applications générales
These highly purified microfibre cloths allow streak-free cleaning of all smooth and polished surfaces such as ceramic, stainless steel, glass and mirror surfaces, surfaces of electronical devices, and monitors.
  • Made of polyester/polyamid (80:20)
  • For multiple use
  • For use in cleanrooms with purity class C/D or ISO class 7/8
Weight: 48 g