Imprimer
Portable refractometers for sea water, PAL series
Fournisseur: Atago
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
4406 4407 4408
635-0217EA 339 EUR
635-0217 635-0218 635-0219
Portable refractometers for sea water, PAL series
Refractometers Handheld Refractometers
These pocket refractometers are designed to deliver rapid results.
Certifications: IP 65 allows rinsing off the sample with running water.
Informations de livraison: Supplied with two AAA batteries.