Imprimer
Nitrogen receivers (schlenk tubes)
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
03370822 03370649
LENZ03370822EA 113 EUR
LENZ03370822 LENZ03370649
Nitrogen receivers (schlenk tubes)
Ballons Flacons pour azote liquide
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With 14/23 standard ground sockets/cones and PTFE needle-valve stopcock with tube connector.