Imprimer
Beakers, with handle, PP, Corning®
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1015P-3L 1015P-1L 1015P-250 1015P-2L 1015P-500
CORN1015P-3LEA 181 EUR
CORN1015P-3L CORN1015P-1L CORN1015P-250 CORN1015P-2L CORN1015P-500
Beakers, with handle, PP, Corning®
Béchers
PP, translucent.
  • Smooth surfaces for easy and fast cleaning
  • Printed with blue graduations
  • With handle and spout
Ideal for general laboratory use and a wide variety of applications.