Imprimer
Pressure relief valves, Upchurch Scientific®
Fournisseur: UPCHURCH SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
P-612 U-455 U-456
UPCHP-612EA 222 EUR
UPCHP-612 UPCHU-455 UPCHU-456
Pressure relief valves, Upchurch Scientific®
Vannes Clapets anti-retour
Pressure relief valves are ideal for preventing system over-pressurisation. These products protect system components by diverting fluid flow automatically when inline pressure exceeds the set limit.
Informations de livraison: Supplied with flangeless fittings. The U-456 assembly is additionally supplied with wrenches for tightening.