Imprimer
Casques de sécurité, uvex pheos E-S-WR
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
9770.131 9770.832 9770.031 9770.431
111-2335EA 21.3 EUR
111-2335 111-2336 111-2334 UVEX9770.431
Casques de sécurité, uvex pheos E-S-WR
Casques Casques de sécurité
The lightweight uvex pheos safety helmets feature a sporty design and offer maximum wearer comfort. The sturdy, rigid helmet shell also easily withstands pressure from the side, and fulfills all safety related requirements for construction or industrial work.
  • Can quickly and easily be adapted to the user's individual needs - visor, safety spectacles, ear defenders or head torch
  • Suspension harness with wheel ratchet for variable width adjustment
  • Six-point textile band attachment ensures optimal fit and comfort
  • With slot on the front for helmet lights and side Euroslot adapters (30 mm) for attaching earmuffs and the uvex pheos visor system
Utilisable par les électriciens, ce casque à coquille est intégralement fermé.
Taille: 520-610 mm
Certifications: EN 397 and requirements for very low temperatures (–30 °C) and molten metal (MM) splash; EN 50365 (1000 V AC).