Imprimer
Alkaline cleaner, TICKOPUR R 33
Fournisseur: BANDELIN-ELECTRONIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
831 883
142-1911EA 102 EUR
142-1911 142-1913
Alkaline cleaner, TICKOPUR R 33
Produits nettoyants Détergents et Nettoyants de Surfaces
Universal cleaner is a corrosion protectant for service, industry, technology and laboratory with gentle cleaning. It can be used to clean glass, metal, ceramics, plastic, rubber and removes grease, oils, wax, silicone films, protein, ink residue, etc.
  • Mildly alkaline
  • Dosage 1 to 5%