Imprimer
Glass tubings, DURAN® borosilicate glass
Fournisseur: Schott AG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
DURAN®
SCOR1027000EA 1580 EUR
SCOR1027000 SCOR1027001 SCOR1027006 SCOR1027005 SCOR1026628 SCOR1026142 SCOR1026141 SCOR1026144 SCOR1026139 SCOR1026138 SCOR1026490 SCOR1225058 SCOR1027020 SCOR1026131 SCOR1027022 SCOR1027021 SCOR1027024 SCOR1026133 SCOR1027023 SCOR1026014 SCOR1026135 SCOR1027026 SCOR1026134 SCOR1027025 SCOR1027028 SCOR1027027 SCOR1026136 SCOR1027019 SCOR1026127 SCOR1027018 SCOR1026646 SCOR1026129 SCOR1139886 SCOR1027011 SCOR1027010 SCOR1026122 SCOR1027013 SCOR1027012 SCOR1027015 SCOR1026123 SCOR1027014 SCOR1027017 SCOR1026125 SCOR1027016 SCOR1027008 SCOR1027007 SCOR1026999 SCOR1027009 SCOR1026718
Glass tubings, DURAN® borosilicate glass
Tuyaux Transfer Tubing
Borosilicate glass acc. to ISO 3585, all-purpose glass for apparatus for the chemical industry, for pipelines and laboratory.
  • High transmittance
  • Electrically highly insulating
  • High chemical resistance
  • High hydrolytic resistance
  • High resistance to thermal shock
  • High temperature resistance
Tube length ~1500 mm.