Imprimer

Agarose Molecular_Biology Type 1, Millipore®

Marques : Merck Millipore Calbiochem
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

121853-100 121853-500GM
121853-100EA 374 EUR
121853-100 121853-500GM
Agarose Molecular_Biology Type 1, Millipore®
Agarose

It is suitable for a wide range of nucleic acid electrophoresis and immunoelectrophoresis applications.


Synonyms: Agarose 10; Agarose 15; Agarose 25; Agarose IEF.

Point de fusion: 87…89 °C
Température de stockage: Température ambiante
Numéro MDL: MFCD00081294
Numéro CAS: 9012-36-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM

Commander maintenant

Résultats des tests de spécification

Gel strength ≥1200 g/cm²
Gelling range (1,5%) 36 - 39 °C
EEO (-mr) ≤0,10
Sulphate ≤0,15%
DNase, RNase, protease None detected
Storage Store at 15 to 30 °C, protect from moisture


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR