Imprimer
Cylinders, borosilicate glass, with hexagonal base
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
213910776 213910879 213911472 213911772 213912477 213913679 213914478 213915474 213916376
SCOT213910776EA 120 EUR
SCOT213910776 SCOT213910879 SCOT213911472 SCOT213911772 SCOT213912477 SCOT213913679 SCOT213914478 SCOT213915474 SCOT213916376
Cylinders, borosilicate glass, with hexagonal base
Éprouvettes Éprouvettes graduées
DURAN® borosilicate glass 3.3, clear.
  • Without graduation
  • Hexagonal base
  • Base with three knobs which increase its stability
Certifications: DIN EN ISO 4788.