Imprimer
ESD safety sandals, Velcro® fastening, Bio, JALAS® 6418
Fournisseur: EJENDALS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6418-36 6418-37 6418-38 6418-39 6418-40 6418-41 6418-42 6418-43 6418-44 6418-45 6418-46 6418-47
EJEN6418-36EA 134 EUR
EJEN6418-36 EJEN6418-37 EJEN6418-38 EJEN6418-39 EJEN6418-40 EJEN6418-41 EJEN6418-42 EJEN6418-43 EJEN6418-44 EJEN6418-45 EJEN6418-46 EJEN6418-47
ESD safety sandals, Velcro® fastening, Bio, JALAS® 6418
Chaussures et bottes
Anti-static ESD safety ankle shoes made from textile microfibre with EVA, polyester based conductive thread insole and polyester polyamide lining.
  • Double shock absorption zones in Poron® XRD®
  • With plastic shank insole and PU outsole
  • ProNose toe reinforcement and oil resistant outsole
  • Composite toe protection cap
  • Puncture resistant midsole made from plasma-treated composite (PTC) textile
Mainly used for indoors, all-year use, metal-free requirements for shoes and environments with risk for outsole penetration.
Colour: Black/grey/green
Certifications: EN 61340-5-1 (ESD); EN ISO 20345:2011; S1P, SRC