Eau, OTEC apyrogène, stérile
Marques : AGUETTANTTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s):
Hydrogen oxide
Sterile purified water is ideal for rinsing instruments, sterile equipment and thermosensitive instruments, cleaning clean rooms, preparation of automated washing solutions, realisation of buffer solutions requiring sterile and pyrogen-free water and raw material for pharmaceutical use.
- Sterile and pyrogen-free solution
- Conductivity at 25 °C: 5.0 μS/cm
- Clear and colourless water
Formule:
H₂O Poids moléculaire: 18.02 g/mol Point d'ébullition: 100 Point de fusion: 0 |
Numéro MDL:
MFCD00011332 Numéro CAS: 7732-18-5 Indice Merck: 13,10098 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...