Imprimer
Probe maintenance kits for dissolved oxygen meters, Orion™
Fournisseur: Thermo Orion
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
O80113 080513
662-0692EA 130 EUR
662-0692 664-0499
Probe maintenance kits for dissolved oxygen meters, Orion™
Électrodes Sondes à oxygène dissous
These probe maintenance kits help ensure optimal performance of Orion™ polarographic dissolved oxygen probes.
Informations de livraison: Each probe maintenance kit includes two membrane caps, one electrolyte solution, and a polishing disk.