Imprimer
Embedding system, Technovit® 7100
Fournisseur: HERAEUS KULZER
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
66009903 64708992 64708996 64708955 64709003 64708991
HKUL66009903EA 59 EUR
HKUL66009903 HKUL64708992 HKUL64708996 HKUL64708955 HKUL64709003 HKUL64708991
Embedding system, Technovit® 7100
Tissue Embedders
Technovit 7100 is a plastic embedding system based on HEMA (2-hydroxyethyl methacrylate). The hydrophillic resin is used in medicine, botany, zoology and in the industry for embedding tissues for light microscope studies. The sections can be used for histological staining and enzyme detection.
  • Reproducibility and reliability of the embedding due to the constant, documented quality controls of the individual components
  • Low polymerisation temperature due to Teflon embedding forms
  • Equal hardening of the block, thus uniform and thinnest possible sections
  • In addition to routine staining, enzyme detection is also possible
  • Less toxic due to catalyst made of barbituric acid
  • Decalcification not necessary for haematological illiac crest biopsies
Technovit 7100 transparently polymerises. Uniform thin sections can be made out of the blocks with the rotation microtome. It is not necessary and also not possible to elute the plastic out of the block and the section.
Areas of applications for embeddings and sections include plastics, foils, paper, textiles, biological products and mounts and wires.
Informations de livraison: Contains a basic solution (500 ml), hardner 1 (5×1 g) and hardner 2 (1×40 ml).