Imprimer
Acriflavine hydrochloride, poudre marron rougeâtre
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Danger
0219067510 0219067525 0219067580
ICNA0219067510EA 45.9 EUR
ICNA0219067510 ICNA0219067525 ICNA0219067580
Acriflavine hydrochloride, poudre marron rougeâtre
Acriflavine hydrochloride
Acriflavine Hydrochloride is a mixture of 3,6-diamino-10-methylacridinium chloride and 3,6-diaminoacridine (approximately an 9 to 1 mixture respectively).
  • UV/Visible Absorbance: λ max (meOH) 460 ± 4 nm
  • Soluble in water (1 mg/ml- clear, dark orange solution).
Acriflavine Hydrochloride acts as an intercalating dye and also reported to inhibit mitochondriogenesis. It is an antiseptic and has been used in applications for dual fluorescence analysis of cellular DNA and protein simultaneously.
Formule: C₂₇H₂₈Cl₄N₆
Point de fusion: 260 °C (déc., lit.)
Température de stockage: Température ambiante
Numéro MDL: MFCD00069039
Numéro CAS: 8063-24-9
UN: 3077
ADR: 9,III
Commander maintenant
Résultats des tests de spécification
Identity Test Passes
UV/Vis: l max (meOH) 460±4nm