Imprimer
Yeast hydrolysate enzymatic from brewers yeast, MP Biomedicals
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0210330480 0210330405 0210330425
ICNA0210330480EA 2589 EUR
ICNA0210330480 ICNA0210330405 ICNA0210330425
Yeast hydrolysate enzymatic from brewers yeast, MP Biomedicals
Milieux pour microbiologie Matières premières de microbiologie
Yeast hydrolysate enzymatic is an autolysed yeast extract produced from 100% brewers yeast. It is a concentrated source of soluble protein components, carbohydrates, and B Complex vitamins natural to yeast.
  • Brown powder