Imprimer
L(+)-Histidine 99, powder_crystalline
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): H, (S)-(+)-Histidine, His, H-His-OH
0210195425 0210195410 0210195480 0210195490 0210195491
ICNA0210195425EA 43.2 EUR
ICNA0210195425 ICNA0210195410 ICNA0210195480 ICNA0210195490 ICNA0210195491
L(+)-Histidine 99, powder_crystalline
L(+)-Histidine
L-histidine is an essential amino acid with a basic side chain, and is hydrophilic in character
Precursor of histamine by action of histidine decarboxylase.
It contains an imidazole group in the side chain. Histidine is biosynthesized from ATP, 5-phosphoribosyl-1-pyrophosphate (PRPP), and glutamine. In turn, histidine is degraded to glutamate by histidase, urocanase, and imidazolonepropionase via the formation of urocanate, 4-imidazolone 5-propionate, and N-formiminoglutamate.
L-Histidine has been used to study cultures of the human T-lymphoblastic leukemia cell line MOLT-4 to study modulation of apoptosis. Exogenous histidine has been shown to enhance the biosynthesis of lovastatin by cultured Aspergillus terreus. Histidine has been utilized as a single nitrogen source to probe swarming in Pseudomonas aeruginosa on agar. It is generally used in media preparation.
Soluble 50 mg/ml in 0,5 M HCl.
Formule: C₆H₉N₃O₂
Poids moléculaire: 155.16 g/mol
Température de stockage: Température ambiante
Numéro MDL: MFCD00064315
Numéro CAS: 71-00-1
Indice Merck: 13,04741
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=badfdb36aac0611e5823a7d05e6cba944f6650fa&vid=9e9f9d881a5b3314c7f342312e77e13493901200&prid=web&clid=SAPDAM