Imprimer
NADPH-Na4 98
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): NADPH tetrasodium salt, Sodium (2R,3R,4R,5R)-2-(6-amino-9H-purin-9-yl)-5-((((((((2R,3S,4R,5R)-5-(3-carbamoylpyridin-1(4H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)oxidophosphoryl)oxy)oxidophosphoryl)oxy)methyl)-4-hydroxytetrahydrofuran-3-yl phosphate, Nicotinamide adenine dinucleotide phosphate tetrasodium salt, (reduced form)
0210116725 0210116750 0210116780 0210116783 0210116791
ICNA0210116725EA 126 EUR
ICNA0210116725 ICNA0210116750 ICNA0210116780 ICNA0210116783 ICNA0210116791
NADPH-Na4 98
NADPH-Na4
Prepared by chemical reduction of NADP.
  • White powder
Store at −0 °C, desiccate, protect from light, store under nitrogen
Formule: C₂₁H₂₆N₇Na₄O₁₇P₃
Poids moléculaire: 833.35 g/mol
Numéro MDL: MFCD00036263
Numéro CAS: 2646-71-1