Imprimer

Eau, OTEC apyrogène, distillé stérile

Marques : AGUETTANT
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

600504 600504_COT
AGUE600504EA 43.2 EUR
AGUE600504 AGUE600504_COT
Eau, OTEC apyrogène, distillé stérile
Eau

Used for cleaning of clean rooms, preparation of automated washing solutions, development of buffer solutions, rinsing of instruments, sterile equipment and thermosensitive instruments. Also used as a raw material for the manufacture of medicines.


Certifications: Conforms to ISO 3696 type 3. Compiant with the monograph 01/2009: 0008 conditioned purified water in a container of the European Pharmacopoeia in force.

Formule: H₂O
Numéro MDL: MFCD00011332
Numéro CAS: 7732-18-5

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR