Imprimer
Sample Bags with Inscription Pad
Fournisseur: NEOLAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1-7090 1-7093 1-7094
NEOL1-7090EA 29.4 EUR
NEOL1-7090 NEOL1-7093 NEOL1-7094
Sample Bags with Inscription Pad
Sachets Sachets d'échantillonnage
Transparent bag made of LDPE with zip-lip closure.
  • Suitable for storing or dispatching of samples
  • Can be very quickly and tightly closed by squeezing
  • With inscription pad