Imprimer
Sample vials, with PTFE-lined screw cap, KIMBLE®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SP60940A-12 SP60940A-8 SP60940A-16 SP60940A-24 SP60940A-4
SCERSP60940A-12EA 291 EUR
SCERSP60940A-12 SCERSP60940A-8 SCERSP60940A-16 SCERSP60940A-24 SCERSP60940A-4
Sample vials, with PTFE-lined screw cap, KIMBLE®
Fioles
These vials are made from clear KIMAX® KG-33 borosilicate glass.
  • Closed top white PTFE closures, attached
  • The 16 ml size complies with EPA 600 series methods