Anti-static low weight ESD safety sandals made from PU-coated and split leather with EVA, polyester based conductive thread insole and polyester-polyamide lining.
- Shock absorption zone in Ergothan
- With plastic shank insole and PU outsole
- Puncture resistant midsole made from plasma-treated composite (PTC) textile
- Toe protection cap made from aluminium (200 J)
Mainly used for indoors, environments with risk for outsole penetration and dry environments.
Colour: Black/grey/red
Certifications: EN 61340 (ESD); EN ISO 20345:2011; S1 P, SRC