Imprimer
ESD safety sandals, Velcro® fastening, E-Sport, JALAS® 1605
Fournisseur: EJENDALS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1605-36 1605-37 1605-38 1605-39 1605-40 1605-41 1605-42 1605-43 1605-44 1605-45 1605-46 1605-47
EJEN1605-36EA 83.27 EUR
EJEN1605-36 EJEN1605-37 EJEN1605-38 EJEN1605-39 EJEN1605-40 EJEN1605-41 EJEN1605-42 EJEN1605-43 EJEN1605-44 EJEN1605-45 EJEN1605-46 EJEN1605-47
ESD safety sandals, Velcro® fastening, E-Sport, JALAS® 1605
Chaussures et bottes
Anti-static low weight ESD safety sandals made from PU-coated and split leather with EVA, polyester based conductive thread insole and polyester-polyamide lining.
  • Shock absorption zone in Ergothan
  • With plastic shank insole and PU outsole
  • Puncture resistant midsole made from plasma-treated composite (PTC) textile
  • Toe protection cap made from aluminium (200 J)
Mainly used for indoors, environments with risk for outsole penetration and dry environments.
Colour: Black/grey/red
Certifications: EN 61340 (ESD); EN ISO 20345:2011; S1 P, SRC