Imprimer
Chemical resistant overalls, MICROGARD® 2500 STANDARD, model 111
Fournisseur: Ansell
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
WH25W-00111-02 WH25W-00111-03 WH25W-00111-04 WH25W-00111-05 WH25W-00111-06 WH25W-00111-07
ANSEWH25W-00111-02EA 572 EUR
ANSEWH25W-00111-02 ANSEWH25W-00111-03 ANSEWH25W-00111-04 ANSEWH25W-00111-05 ANSEWH25W-00111-06 ANSEWH25W-00111-07
Chemical resistant overalls, MICROGARD® 2500 STANDARD, model 111
Vêtements à usage unique
These overalls are made from a durable microporous PP laminate with a non woven inner layer. The fabric provides an excellent barrier to chemical spray and infective agents.
  • 3-piece hood is designed for optimum fit with respirators, particularly full face masks
  • Elasticated hood, wrist, waist and ankles
  • 2-way front zip with resealable storm flap
  • Finger loops to prevent sleeve movement when working above the head
  • Ultrasonically welded seams provide a strong liquid and particle barrier
  • Moisture vapour permeable ('breathable') to help reduce the risk of heat stress
  • Low linting - Reduced risk of contamination in critical areas
Typical applications include: Viral contaminated areas (including Avian Influenza), biological protection, emergency medical response, medical research, chemical and pharmaceutical industries, low pressure industrial cleaning, industrial paint spraying, nuclear industry.
Certifications: Type 4-B/5-B/6-B, EN 14126, EN 1073-2, EN 1149-5; Cat. III.