Imprimer
Cut resistant gloves, HyFlex® 11-727
Fournisseur: Ansell
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
11-727/8 11-727/9 11-727/6 11-727/7 11-727/10 11-727/11 11-727/5
ANSE11-727/8EA 172 EUR
ANSE11-727/8 ANSE11-727/9 ANSE11-727/6 ANSE11-727/7 ANSE11-727/10 ANSE11-727/11 ANSE11-727/5
Cut resistant gloves, HyFlex® 11-727
Gants
These knitted seamless liners with INTERCEPT™ technology yarn and PU coated palm combine excellent comfort and mobility with optimal cut protection.
  • Best-in-class cut protection and increased comfort and dexterity
  • Palm coating design improves worker grip and extends glove life in abrasive conditions
  • Uncoated back allows the hand to breath
  • Grey coating hides dirt in environments with dirt exposure
INTERCEPT™ is a specialised knitting technology which blends engineered, synthetic and natural fibres into high performance yarns. This provides high-cut protection with exceptional comfort and dexterity.
Ideal for assembling white goods, general maintenance, glass manufacturing, medical device manufacturing, plastics injection and moulding, and pre-assembling. Also suitable for handling sharp-edged objects and for cutting small dry or lightly oiled parts.
Certifications: EN 388:2016 (4342B); Cat. II