Imprimer
McFarland turbidity standards, Remel™
Marques : THERMO FISHER DIAGNOSTICS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
R20411 R20412 R20413 R20414 R20421 R20410
OXOIR20411EA 77.8 EUR
OXOIR20411 OXOIR20412 OXOIR20413 OXOIR20414 OXOIR20421 OXOIR20410
McFarland turbidity standards, Remel™
Étalons Étalons de turbidité
These McFarland equivalence turbidity standard is packed in plastic case with a visual comparision card. Tube size of 15×103 mm.
  • Fits in most spectrometers
  • Polystyrene technology allows longer shelf life
Informations de livraison: The turbidity standard set includes 1 each of 0,5, 1,0, 2,0, 3,0 and 4,0.