Imprimer
Échantillonneurs à usage unique, DispoLadle
Fournisseur: BURKLE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5393-6640 5393-6641 5393-6650 5393-6651
BURK5393-6640EA 140 EUR
BURK5393-6640 BURK5393-6641 BURK5393-6650 BURK5393-6651
Échantillonneurs à usage unique, DispoLadle
Échantillonneurs Échantillonneurs à usage unique
DispoLadle is a sampling scoop specially designed for disposable use. The scaled scoop with embossing can be used for measuring. It shows contents in increments of 10 ml.
  • Ergonomic handle and a spout on both sides for right or left-hand operation
  • Clean room manufactured, individually packaged
  • Optionally sterilised by gamma rays