Imprimer
Labels for BBP33 printers, type B-499
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
143001 361682 142997 142995 143002 143049 143066 143015 143016 143017 143018 143011 361672 361681 361663 361694 361664 361665 361666 361679 153086 153095 153096 153093 153089 153088 153087 153092 153091 153085 153090 153094
BRDY143001EA 172 EUR
BRDY143001 BRDY361682 BRDY142997 BRDY142995 BRDY143002 BRDY143049 BRDY143066 BRDY143015 BRDY143016 BRDY143017 BRDY143018 BRDY143011 BRDY361672 BRDY361681 BRDY361663 BRDY361694 BRDY361664 BRDY361665 BRDY361666 BRDY361679 BRDY153086 BRDY153095 BRDY153096 BRDY153093 BRDY153089 BRDY153088 BRDY153087 BRDY153092 BRDY153091 BRDY153085 BRDY153090 BRDY153094
Labels for BBP33 printers, type B-499
Étiquettes
These nylon cloth labels are used for electrical and automation identification, inside-panel component labelling, wire and cable marking.