Imprimer
Labels for BBP33 printers, type B-492, FreezerBondz™
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
143082 143000 142917 143051 143063 143067 143100 142919 142918 134102 134096 307018
BRDY143082EA 204 EUR
BRDY143082 BRDY143000 BRDY142917 BRDY143051 BRDY143063 BRDY143067 BRDY143100 BRDY142919 BRDY142918 BRDY134102 BRDY134096 BRDY307018
Labels for BBP33 printers, type B-492, FreezerBondz™
Étiquettes
FreezerBondz™ material is a low-profile label used on frozen surfaces or on room-temperature surfaces that will eventually be stored in liquid nitrogen (–196 °C). Opaque enough to cover legally protected patient information for compliance with HIPAA requirements.
  • Ideal for labelling frosted, frozen or room temperature vials (with overlap)
  • Good solvent resistance when printed with R-6400 series ribbon