Imprimer
Labels for BBP33 printers, type B-490, FreezerBondz
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
142938 142943 142940 142894 142939 142942 143089 142941 142893 142895 142896
BRDY142938EA 134 EUR
BRDY142938 BRDY142943 BRDY142940 BRDY142894 BRDY142939 BRDY142942 BRDY143089 BRDY142941 BRDY142893 BRDY142895 BRDY142896
Labels for BBP33 printers, type B-490, FreezerBondz
Étiquettes
Freezerbondz™ polyester labels can be used to label frozen surfaces (with overlap) including glass and polypropylene stored in liquid nitrogen.