Imprimer
Real-Time PCR kit for the detection of the allergen hazel-nut in food samples
Marques : Q BIOANALYTIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
QB-RT-19-1000 QB-RT-19-250 QB-RT-19-50
QBIAQB-RT-19-1000EA 0 EUR
QBIAQB-RT-19-1000 QBIAQB-RT-19-250 QBIAQB-RT-19-50
Real-Time PCR kit for the detection of the allergen hazel-nut in food samples
Tests microbiologiques Tests PCR
Hazel-nut is allergenic for a considerable number of individuals. Using DNA based tests also processed food samples might be tested. Our Real-Time PCR test exhibits the required limit of detection laid down in the codex alimentarius. The Real-Time PCR kit has a limit of detection of ≤ 20 DNA-copies. The limit of detection can be influenced by sample matrix, level of processing, DNA-preparation and DNA-content.
  • TaqMan probes
  • Taget in FAM channel
  • Contains UNG to protect from re-amplification
  • Positive DNA and negative control included