Imprimer
Real-Time PCR kit for detection of toxic Vibrio parahaemolyticus
Marques : Q BIOANALYTIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
QB-RTI-50-1000 QBRTI-50-10SMP QB-RTI-50-250 QB-RTI-50-50
710-1773EA 0 EUR
710-1773 710-1774 710-1775 710-1776
Real-Time PCR kit for detection of toxic Vibrio parahaemolyticus
Tests microbiologiques Tests PCR
Vibrio parahaemolyticus tox gene (tdh/trh): Food safety experts like the German DGHM recommend to test warm water fish for Vibrio parahaemolyticus. With this Real-Time PCR test you have a very versatile method for testing whether or not the present strain has the ability to produce toxins (tdh/trh). You can use our Real-Time PCR Test for Vibrio parahaemolyticus for testing for the presence of the species before. You can use this test in combination with an appropriate pre- enrichment in alkaline peptone-water or you can confirm bacterial colonies on agar plates.
  • TaqMan probes
  • Contains internal amplification control in HEX channel
  • Taget in FAM channel
  • Contains UNG to protect from re-amplification
  • Positive DNA and negative control included
Certifications: DIN EN ISO 20838 conform detection in food, feed and water samples from pre-enrichment cultures or for confirmation of bacterial colonies.