Imprimer
Anion exchange chromatography columns, TSK-GEL®, STAT
Fournisseur: TOSOH Bioscience
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0021961 0021960 0021962
TOYO0021961EA 1970 EUR
TOYO0021961 TOYO0021960 554-5787
Anion exchange chromatography columns, TSK-GEL®, STAT
Colonnes pour chromatographie
TSK-Gel® STAT are polymer based columns for high resolution protein separations like nucleic acids, mAb variants, PEGylated protein and protein aggregates.
  • Cross-linked hydrophilic polymer
  • Non porous
DNA STAT column are designed for high resolution DNA separations.