Imprimer
Rods, borosilicate glass
Fournisseur: HILGENBERG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
7347701 7347720 7347730 7347710
HILG7347701EA 1080 EUR
HILG7347701 HILG7347720 HILG7347730 HILG7347710
Rods, borosilicate glass
Tiges solides
Glass rods are made of borosilicate glass 3.3.
  • Very good chemical resistance
  • Very good temperature resistance
  • Length: 100 mm
With ends cut or fixing of samples and filling of mark-tubes.