Imprimer
Steel ground protection systems
Fournisseur: CEMO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
7401 7402 10617 7403 7404 7405 10618 7406
CEMG7401EA 889 EUR
CEMG7401 CEMG7402 CEMG10617 CEMG7403 CEMG7404 CEMG7405 CEMG10618 CEMG7406
Steel ground protection systems
Matériaux absorbants
These ground protection systems ensures safe handling of water-polluting as well as flammable substances - previously classed as AI*, AII, AIII and B.
  • Storage of water-polluting and flammable substances
  • Painted steel RAL 5002, ultramarine blue
  • Hot-dip galvanised gratings
Modular matched elements enable the lining of any large surfaces as a storage or transfilling station.