Imprimer
Tubing for rotarus® peristaltic pumps, Tygon® Chemical
Fournisseur: HIRSCHMANN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
95215013 95215018 95215016 95215015
HIRS95215013EA 497 EUR
HIRS95215013 HIRS95215018 HIRS95215016 HIRS95215015
Tubing for rotarus® peristaltic pumps, Tygon® Chemical
Tuyaux Transfer Tubing
These tubes are used in chemical application for its very high chemical resistance to acids, bases, salts, ketones, alcohols.
  • Temperature resistance from –60 to 74 °C
  • Hardness (Shore durometer A): 61