Imprimer
Mobile sumps for storage of hazardous materials, GRP
Fournisseur: CEMO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
7870 7834
CEMG7870EA 748 EUR
CEMG7870 CEMG7834
Mobile sumps for storage of hazardous materials, GRP
Chariots
The mobile sumps are applicable for the storage and transport of environmentally hazardous substances.
  • Sumps contains hot-dip galvanised grating and chassis with collision protection
  • It has two steering and two fixed castors and a pushing bar for trouble-free steering
  • With general construction inspection approval number Z-40.12-227
The thermosetting plastic base structure in conjugation with the glass reinforcement provide an advantageous fire behaviour and high chemical reistance.