Imprimer
Sterilisation Boxes for Pipettes
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
28920 28925 28930 28935
BRND28920EA 62.5 EUR
BRND28920 BRND28925 BRND28930 BRND28935
Sterilisation Boxes for Pipettes
Boîtes Boîtes de stérilisation pour pipettes
These pipette boxes made of aluminum are ideal for the storage and sterilisation of pipettes with different lengths.
  • With inner silicone coating
  • Very good temperature resistance
  • Very good chemical resistance