Imprimer
Réfractomètres portables, série MASTER
Fournisseur: Atago
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2354 2351 2352 2353 2383 2382 2381 2971 2972 2973 2991 2992 2993
635-0691EA 305 EUR
635-0691 635-0688 635-0689 635-0690 635-0700 635-0699 635-0698 635-0667 635-0668 635-0669 635-0673 635-0674 635-0675
Réfractomètres portables, série MASTER
Réfractomètres Réfractomètres portables
The MASTER series of handheld refractometers provides a range of easy to use instruments for a wide variety of applications.
  • MASTER-M: Standard model, clear scale, only two drops required
  • MASTER-T: Automated temperature compensation from 10 to 30 °C
  • MASTER-α: Automated temperature compensation (IP 65 except eyepiece), dust-tight and protected against water jets
  • MASTER-P: Designed with a plastic body for resistance to salty and acidic samples
The design has an ergonomic grip and a prism housing that allows for the sample to cover the prism without having to lift the daylight plate.