Imprimer
Screw caps for screw neck vials, N 18 and N 20
Fournisseur: MACHEREY-NAGEL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
702181 702115
MANA702181EA 92.8 EUR
MANA702181 MANA702115
Screw caps for screw neck vials, N 18 and N 20
Capuchons Bouchons pour flacons de chromatographie
The suitable screw closures have bonded septa.
  • Septa are firmly connected with the cap (bonded)
  • High quality silicone/PTFE septum materials