Imprimer
Thermal transfer printer labels, type B-481, StainerBondz
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
108241 804785 804784 116385 108242 804787
BRDY108241EA 384 EUR
BRDY108241 BRDY804785 BRDY804784 BRDY116385 BRDY108242 BRDY804787
Thermal transfer printer labels, type B-481, StainerBondz
Étiquettes
These labels coupled with R-6400 series ribbon will remain resistant when exposed to xylene and other chemicals that are commonly used in the slide staining process.
  • No print removal and label buckling
  • No discolouration that would cause barcodes to be unreadable
  • Stays adhered to the slide