Imprimer
Human albumin (from serum), MP Biomedicals
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
08823030 08823031 08823032 08823034
ICNA08823030EA 80.4 EUR
ICNA08823030 ICNA08823031 ICNA08823032 ICNA08823034
Human albumin (from serum), MP Biomedicals
Protéines et Peptides
Albumins are soluble monomeric proteins found in the body fluids and tissues of animals and in some plant seeds. It functions as a carrier protein for steroids, fatty acids, and thyroid hormones.
  • Isoelectric point (pI): 5,3 (Lit.)
It plays a vital in regulating the colloidal osmotic pressures of blood.
It was prepared using Cohn method IV. The material is non-reactive for HbsAg, Anti-HCV 3,0, Anti-HIV-1/2 and STS (RPR). This material is also negative for HIV-1 and HCV by PCR.