Imprimer
Corning® Cryogenic Storage Boxes for Corning® 2 ml Polypropylene Cryogenic Vial, with 1D and 2D Barcodes
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8673 8674
479-1889EA 148 EUR
479-1889 479-1890
Corning® Cryogenic Storage Boxes for Corning® 2 ml Polypropylene Cryogenic Vial, with 1D and 2D Barcodes
Boîtes Boîtes pour cryotubes
Reusable polycarbonate boxes.
  • For 1 and 2 ml vials
  • Ridges on sides of box make them easier to grip while wearing gloves