Imprimer
Standard fire fighting equipment signs
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
817047 817206 816890 817049 816881 253600 817683 816900 816895 816899 817054 817690
BRDY817047EA 11 EUR
BRDY817047 BRDY817206 BRDY816890 BRDY817049 BRDY816881 BRDY253600 BRDY817683 BRDY816900 BRDY816895 BRDY816899 BRDY817054 BRDY817690
Standard fire fighting equipment signs
Panneaux
A range of signs to clearly mark the location of fire extinguishers and other fire fighting equipment as well as fire alarm points.
  • Used for a wide range of applications
  • Resistance to most chemical products and solvents
  • Non-adhesive
  • Thickness: 1,5 mm
  • Different sizes
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable